Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2D Mouse, Unlabeled, Clone: 9, BD

Catalog No. BDB610774 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610774 50 μg
BDB610775 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610774 Supplier BD Biosciences Supplier No. 610774
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEF2D is a member of the MEF2 (Myocyte-specific Enhancer-binding Factor 2) transcription factor family. Members of this family (MEF2A, B, C, & D) contain several characteristic motifs that include a MEF2 domain and a MADS box. MEF2 proteins bind the DNA sequence, 5′- (C/T)T(A/T)(A/T)AAATA(A/G)-3′, and binding occurs primarily, although not exclusively, in muscle cells. MEF2D gene expression is ubiquitous and the mRNA is alternatively spliced. This results in multiple protein isoforms that heterodimerize with other family members. In addition to the MEF2 and MADS domains, MEF2D has a glutamine-/proline-rich region, amino acids 365-404, and a basic region, in the MADS domain near the N-terminus. The glutamine-/proline-rich region is involved in transactivation, while the basic region mediates sequence-specific DNA binding. MEF2D displays 75% similarity to hMEF2A and hMEF2C. Myogenin gene transcription requires MEF2D, indicating that MEF2D is involved in myogenesis and muscle cell differentiation. MEF2D has been reported to have a calculated molecular weight of 56kDa, but can be observed migrating in a range of 56-70kDa.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2D
Applications Western Blot
Classification Monoclonal
Clone 9
Concentration 250μg/mL
Conjugate Unconjugated
Description Myocyte-specific Enhancer-binding Factor 2
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse MEF2D aa. 346-511
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.