Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meis 1/2/3 Mouse anti-Human, Mouse, Clone: 9.2.7, MilliporeSigma™

Catalog No. 05779MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-779-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-779-MI Supplier MilliporeSigma Supplier No. 05779
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Meis 1/2/3 with Anti-Meis 1/2/3 Antibody, clone 9.2.7 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Meis 1/2/3
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9.2.7
Conjugate Unconjugated
Formulation mouse ascites IgG1 containing 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_170677
Gene Symbols MEIS2, MGC2820, MRG1, HsT18361
Host Species Mouse
Immunogen MBP-conjugated fusion protein corresponding to amino acids 60-390 of mouse Meis1
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.