Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07641MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-641-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-641-MI Supplier MilliporeSigma Supplier No. 07641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-MEK1 Antibody is validated for use in immunoprecipitation, western blotting for the detection of MEK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 70% storage buffer (0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide) and 30% glycerol.
Gene Symbols MAP2K1; PRKMK1; MKK1; MAPKK1; MEK1
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 2-18 (PKKKPTPIQLNPAPDGS) of human MEK1.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002755
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.