Learn More
Description
The O24-836 monoclonal antibody specifically binds to the human MEK1 pS218 and MEK2 pS222 phosphorylated sites. MEK1 and MEK2 (MEK1/MEK2) are ∽45 kDa dual-specificity protein kinases that are also known as MAPK/ERK Kinase 1 and 2 and MAP kinase kinases (aka, MAP2K1 and 2, MAPKK1 and 2, or MKK1 and 2). MEK1 and MEK2 activation is dependent upon phosphorylation of certain amino acid residues including serine 218 (pS218) of MEK1 and serine 222 (pS222) of MEK2 by MAP kinase kinase kinases (MAP3Ks), such as by Raf kinases. MEK1/MEK2 can phosphorylate and thus activate the MAP (Mitogen-Activated Protein) kinases, ERK1 and ERK2. The ERK kinases are critically involved in multiple signal transduction pathways induced by growth factors, cytokines, and mitogens. These kinases regulate cellular growth, differentiation, survival, motility and angiogenesis. Dysregulated activation of MEK1/2 is observed in tumor cells. During antibody development, the purified O24-836 monoclonal antibody was found to detect phosphorylated MEK1/MEK2 by Western blot analysis of cellular lysates and by indirect immunofluorescent staining and flow cytometric analysis of fixed and permeabilized cells. This antibody crossreacts with phosphorylated MEK1/MEK2 expressed by mouse cells as tested by Western blot analysis and immunofluorescent staining and flow cytometric analysis.
Specifications
Specifications
| Antigen | MEK1 (pS218)/MEK2 (pS222), PE |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | O24-836 |
| Conjugate | PE |
| Description | MAP2K1/2; MAPKK1/2; MAP kinase kinase 1/2; MAP2K1/2; PRKMK1/2 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Phosphorylated Human MEK1 Peptide |
| Purification Method | Affinity Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.