Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1 (pS298) Mouse anti-Human, Alexa Fluor 488, Clone: J114-64, BD

Catalog No. BDB560192 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560192 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560192 Supplier BD Biosciences Supplier No. 560192

Mouse Monoclonal Antibody

MEK (Map/Erk Kinase) 1 and 2 are serine/threonine kinases, also known as MAP kinase kinases (MAP2K1 and 2, MAPKK1 and 2, or MKK1 and 2). They activate the MAP (Mitogen-Activated Protein) kinases, also known as ERKs (Extracellular signal Regulated Kinases), which are critical kinases in multiple signal transduction pathways that regulate cell growth and differentiation. Activation of MEK 1 and 2 is dependent upon phosphorylation of serines 218 and/or 222 by activated MAP kinase kinase kinases (MAP3Ks), such as the Raf isoforms. Hormones, growth and differentiating factors, or tumor promoters induce Raf activation via activation of Ras proteins. Alternatively, cellular adhesion can lead to phosphorylation of MEK1 at serine 298 (S298), mediated by p21-activated kinase (PAK). The S298-phosphorylated MEK1 has an enhanced capacity to interact with Raf, resulting in MEK1 activation. The J114-64 monoclonal antibody recognizes the phosphorylated S298 of MEK1.

Host Species: Mouse
Clone: J114-64
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human MEK1 Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK1 (pS298)
Applications Flow Cytometry
Classification Monoclonal
Clone J114-64
Conjugate Alexa Fluor 488
Description MAP2K1, MAPKK1, MKK1, MP2K1, PPKMK1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human MEK1 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.