Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK5 Mouse, Unlabeled, Clone: 21, BD

Catalog No. BDB610956 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610956 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610956 Supplier BD Biosciences Supplier No. 610956

Mouse Monoclonal Antibody

The mitogen-activated protein (MAP) kinase cascade participates in the Ras signal transduction pathway. The MAP kinase cascade consists of MEK kinase (Raf-1 or B-Raf), MAP/ERK kinase (MEK1 or MEK2), and extracellular-regulated protein kinase (ERK1 or ERK2). Raf becomes activated following its interaction with Ras-GTP (activated Ras). Via phosphorylation reactions, Raf activates MEK which, in turn, activates ERK. MEK5, also known as MAP Kinase 5, is a 50kDa protein that is 40% identical to MEK1 and MEK2. Alternative splicing results in two MEK5 isoforms: α (50kDa) and β (40kDa). MEK5β has been reported to be ubiquitously expressed while MEK5α is expressed primarily in the liver and brain. The N-terminal amino acid sequence of MEK5α resembles sequences found in actin cytoskeletal proteins, indicating that MEK5α may associate with the cytoskeleton. However, the role of MEK5 in the MAP kinase pathway and its substrate remain to be identified.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK5
Applications Western Blot
Classification Monoclonal
Clone 21
Concentration 250μg/mL
Conjugate Unconjugated
Description MAP Kinase-5
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MEK5 aa. 13-188
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.