Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MEK7 (Ser271, Thr275), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA128418
Description
PA1-28418 detects MEK7 from human samples.PA1-28418 has been successfully used in Western blot applications.The PA1-28418 immunogen is: Synthetic peptide (Human) derived from a region of human MEK 7 that contains serine 271 and threonine 275. The sequence is conserved in mouse.
Specifications
Phospho-MEK7/MAP2K7 pSer271 + pThr275 | |
Polyclonal | |
Unconjugated | |
O14733, Q8CE90 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5609 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Purified |
Western Blot | |
1mg/mL | |
MAP2K7 | |
MAP2K7 | |
Synthetic peptide (Human) derived from a region of human MEK 7 that contains serine 271 and threonine 275. The sequence is conserved in mouse. | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction