Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEK1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515100
Description
This antibody is predicted to react with mouse and Xenopus based on sequence homology.
MAP2K4 (SEK1) is a dual specificity protein kinase that belongs to the Ser/Thr protein kinase family. This kinase is a direct activator of MAP kinases in response to various environmental stresses or mitogenic stimuli. It has been shown to activate MAPK8/JNK1, MAPK9/JNK2, and MAPK14/p38, but not MAPK1/ERK2 or MAPK3/ERK3. MAP2K4 is phosphorylated and activated by MAP3K1/MEK.
Specifications
SEK1 | |
Polyclonal | |
MAP2K4 | |
Dual specificity mitogen-activated protein kinase kinase 4; MAP kinase kinase 4; JNK-activating kinase 1; c-Jun N-terminal kinase kinase 1; JNKK; SAPK/ERK kinase 1; SEK1; MAP2K4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6416 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P45985 | |
MAP2K4 | |
KLH conjugated synthetic peptide between 235-264 amino acids from human MEK4 (MAP2K4) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction