Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanocyte Stimulating Hormone α Sheep anti-Mammals, Polyclonal, MilliporeSigma™

Catalog No. AB5087MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5087MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5087MI Supplier MilliporeSigma Supplier No. AB5087
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Anti-Melanocyte Stimulating Hormone Antibody, α detects level of Melanocyte Stimulating Hormone and has been published and validated for use in immunohistochemistry, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melanocyte Stimulating Hormone α
Applications Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Sheep Serum. Liquid. Contains no preservative.
Gene Symbols POMC; LPH; NPP; POC; ACTH; CLIP
Host Species Sheep
Immunogen MSH conjugated with bovine thyroglobulin.
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000939.2; NM_001035256.1
Target Species Mammalia
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.