Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MELK Mouse anti-Human, Clone: 6C1.3, MilliporeSigma™

Catalog No. MAB4331MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4331MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4331MI Supplier MilliporeSigma Supplier No. MAB4331
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-MELK Antibody, clone 6C1.3 is validated for use in western blotting for the detection of MELK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MELK
Applications Western Blot
Classification Monoclonal
Clone 6C1.3
Conjugate Unconjugated
Formulation Purified ascites. Liquid in PBS, pH 7.0. Contains 0.1% sodium azide.
Gene Accession No. NM_014791.2
Gene Symbols MELK, hMELK, HPK38, KIAA0175, hPK38, OTTHUMP00000046113, EC 2.7.11.1
Host Species Mouse
Immunogen MELK Recombinant protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.