Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melusin Mouse, Unlabeled, Clone: 42, BD

Catalog No. BDB611894 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611894 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611894 Supplier BD Biosciences Supplier No. 611894

Mouse Monoclonal Antibody

Integrins are membrane receptors that mediate cell-cell or cell-matrix adhesion. All integrins are transmembrane heterodimers composed of α and β subunits that are connected to the cytoskeleton. In mammals, at least 17 α subunits and 8 β subunits have been identified, and these proteins can heterodimerize to form at least 22 different receptors. The β1 subgroup of the integrin receptors consists of at least 6 different dimer combinations. A variety of signal transduction proteins have been shown to bind the cytoplamic domain of β1 integrins. These include melusin, ILK, ICAP, and RACK1. Melusin is expressed preferentially in muscle and heart, and contains putative SH3 domain binding motifs in the N-terminal region, two putative SH2 binding sites, and a C-terminal acidic amino acid stretch (CAAS) similar to the calcium binding proteins, calreticulin and calsequestrin. In muscle, melusin protein is localized in two rows flanking α-actinin at Z-lines, and melusin mRNA is upregulated during neonatal development. Differentiation of C2C12 murine myogenic cell line by serum starvation also upregulates melusin protein and mRNA. Thus, melusin may be an important signal transducer for β1 integrins during muscle development.

Host Species: Mouse
Clone: 42
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Mouse Melusin aa. 61-258

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Melusin
Applications Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Melusin aa. 61-258
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.