Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mesoderm posterior 2 homolog (mouse), Mouse, Clone: 1C8, Abnova™

Catalog No. 89001397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-397 Supplier Abnova Corporation Supplier No. H00145873M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MESP2.

This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02). [provided by RefSeq

Sequence: AQSPPPQSLLGHDHWIFAQGWGWAGHWDSTSPASSSDSSGSCPCDGARGLPQPQPPSCSSRAAEAAATTPRRARTGPAGGQRQSASEREKLRMRTLARALHELRRFLPPSLAPAGQSLTKIETL

Specifications

Antigen mesoderm posterior 2 homolog (mouse)
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 1C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MESP2.
Formulation PBS with no preservative; pH 7.4
Gene MESP2
Gene Accession No. XM_085261
Gene Alias SCDO2/bHLHc6
Gene Symbols MESP2
Host Species Mouse
Immunogen MESP2 (XP_085261, 2 a.a. ∼ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 145873
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.