Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Met/HGFR Mouse anti-Human, Mouse, Clone: 6AT1877, MilliporeSigma™

Catalog No. MAB3731MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3731MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3731MI Supplier MilliporeSigma Supplier No. MAB3731
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Met/HGFR Antibody, phospho-specific (Tyr1234/1235), clone 6AT1877 detects level of Met/HGFR and has been published and validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Met/HGFR
Applications ELISA
Classification Monoclonal
Clone 6AT1877
Conjugate Unconjugated
Formulation Protein G purified immunoglobulin. Liquid in PBS containing 0.09% sodium azide.
Gene Accession No. NM_000245.2
Gene Symbols MET, AUTS9, c-Met, RCCP2, HGFR, EC 2.7.10.1
Host Species Mouse
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Tyr1234/1235 of human Met.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.