Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metabotropic Glutamate Receptor 2/3 Rabbit anti-Monkey, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1553MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1553MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1553MI Supplier MilliporeSigma Supplier No. AB1553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Metabotropic Glutamate Receptor 2/3 Antibody detects level of Metabotropic Glutamate Receptor 2/3 and has been published and validated for use in immunohistochemistry (paraffin), immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Metabotropic Glutamate Receptor 2/3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified using the immunogen peptide. Lyophilized from PBS containing 1% BSA.
Gene Symbols GRM2; mGlu2; GPRC1B; MGLUR2; mGluR2; GLUR2
Host Species Rabbit
Immunogen Carboxy terminus peptide of rat mGluR2 conjugated to BSA with glutaraldehyde (NGREVVDSTTSSL).
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000839.2
Target Species Monkey, Rat
Content And Storage Lyophilized: 2-8°C or −20°C. Reconstituted: −20°C or −80°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.