Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metadherin Mouse anti-Human, Clone: 2F11C3, MilliporeSigma™

Catalog No. MABN699MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN699MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN699MI Supplier MilliporeSigma Supplier No. MABN699
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Metadherin using this mouse monoclonal antibody, Anti-Metadherin Antibody, clone 2F11C3 validated for use in western blotting, immunohistochemistry and Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Metadherin
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2F11C3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in PBS containing up to 0.1% sodium azide and 50% glycerol.
Gene Accession No. NP_848927
Gene Symbols 3D3, AEG1, LYRIC, MTDH
Host Species Mouse
Immunogen Purified protein fragment of Human Metadherin expressed in E. coli.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.