Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Metapneumovirus 75.1 Mouse anti-Human, Clone: 2D7, MilliporeSigma™

Catalog No. MAB8515MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8515MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8515MI Supplier MilliporeSigma Supplier No. MAB8515
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Human Metapneumovirus 75.1 Antibody, clone 2D7 is validated for use in ELISA, immunofluorescence for the detection of Human Metapneumovirus 75.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Metapneumovirus 75.1
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 2D7
Conjugate Unconjugated
Formulation Purified immunoglobulin. 0.02MPhosphate Buffered Saline, 0.25MSodium Chloride, 0.1% Sodium Azide, pH = 7.6
Host Species Mouse
Immunogen MPV CAN98-75
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.