Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-PP2A Mouse anti-Human, Mouse, Yeast (S. cerevisiae), Rat, Pig, Rabbit, Chicken, Drosophila, Clone: 2A10, MilliporeSigma™

Catalog No. 041479MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-147-9MI 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-147-9MI Supplier MilliporeSigma Supplier No. 041479
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-methyl-PP2A Antibody, C subunit, clone 2A10 is validated for use in western blotting for the detection of methyl-PP2A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen methyl-PP2A
Applications Western Blot
Classification Monoclonal
Clone 2A10
Conjugate Unconjugated
Formulation Purified mouse monoclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl and 0.05% sodium azide.
Gene Accession No. NP_002706.1
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide (C-βA-RRTPDYFL-Ome) corresponding to amino acids 302-309 of human PP2A catalytic subunit. Clone 2A10.
Purification Method Protein G purfied
Quantity 150 μg
Primary or Secondary Primary
Target Species Chicken, Drosophila, Human, Mouse, Pig, Rabbit, Rat, S. cerevisiae, Yeast
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.