Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl VCP/p97 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABT286MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT286MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT286MI Supplier MilliporeSigma Supplier No. ABT286

Rabbit Polyclonal Antibody

Detect Valosin-containing protein using this rabbit polyclonal antibody, Anti-methyl VCP/p97 Antibody (Lys315) validated for use in western blotting, immunoprecipitation and Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen methyl VCP/p97 (Lys315)
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide and 30% glycerol.
Gene Symbols VCP
Host Species Rabbit
Immunogen Linear peptide corresponding to human VCP/p97 methylated at Lys315.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_009057
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.