Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017129 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-129 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-129 Supplier Abnova Corporation Supplier No. H00055798B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human METTL2 protein.

This gene is a member of a family of methyltransferases that share homology with, but are distinct from, the UbiE family of methyltransferases. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq

Sequence: MPVDYEINAHKYWNDFYKIHENGFFKDRHWLFTEFPELAPSQNQNHLKDWFLENKSEVCECRNNEDGPGLIMEEQHKCSSKSLEHKTQTPPVEENVTQKISDLEICADEFPGSSATYRILEVGCGVGNTVFPILQTNNDPGLFVYCCDFSSTAIELVQTNSEYDPSRCFAFVHDLCDEEKSYPVPKGSLDIIILIFVLSAIVPDKMQKAINRLSRLLKPGGMVLLRDYGRYDMAQLRFKKGQCLSGNFYVRGDGTRVYFFTQEELDTLFTTAGLEKVQNLVDRRLQVNRGKQLTMYRVWIQCKYCKPLLSSTS

Specifications

Antigen METTL2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human METTL2 protein.
Formulation PBS with no preservative; pH 7.4
Gene METTL2B
Gene Accession No. BC107586.1
Gene Alias FLJ11350/FLJ12760/METL/METTL2/METTL2A/PSENIP1
Gene Symbols METTL2B
Host Species Mouse
Immunogen METTL2 (AAI07587.1, 1 a.a. ∼ 313 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55798
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.