Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

exoribonuclease 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-602 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-602 Supplier Abnova Corporation Supplier No. H00112479B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MGC16943 protein.

Sequence: MATKRLARQLGLIRRKSIAPANGNLGRSKSKQLFDYLIVIDFESTCWNDGKHHHSQEIIEFPAVLLNTSTGQIDSEFQAYVQPQEHPILSEFCMELTGIKQAQVDEGVPLKICLSQFCKWIHKIQQQKNIIFATGISEPSASEVKLCAFVTWSDWDLGVCLEYECKRKQLLKPVFLNSWIDLRATYKLFYRRKPKGLSGALQEVGIEFSGREHSGLDDSRNTALLAWKMIRDGCVMKITRSLNKGPFLLPSWTWNSDLASGDQHAFLKQEFGCGTYRTLLQKPNMSKQEKGNILWLTMVWLSLACLQRKNYNDCMLNTASQTVTTEKF

Specifications

Antigen exoribonuclease 2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MGC16943 protein.
Formulation PBS with no preservative; pH 7.4
Gene ERI2
Gene Accession No. NM_080663.1
Gene Alias EXOD1/KIAA1504/MGC16943
Gene Symbols ERI2
Host Species Mouse
Immunogen MGC16943 (NP_542394.1, 1 a.a. ∼ 328 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112479
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.