Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transcriptional adaptor 2 (ADA2 homolog, yeast)-beta, Mouse, Clone: 1C8, Abnova™

Catalog No. 89014344 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-344 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-344 Supplier Abnova Corporation Supplier No. H00093624M08
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MGC21874.

Sequence: AELGKKYCVYCLAEVSPLRFRCTECQDIELCPECFSAGAEIGHHRRYHGYQLVDGGRFTLWGPEAEGGWTSREEQLLLDAIEQFGFGNWEDMAAHVGASRTPQEVMEHY

Specifications

Antigen transcriptional adaptor 2 (ADA2 homolog, yeast)-beta
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MGC21874.
Formulation PBS with no preservative; pH 7.4
Gene TADA2B
Gene Accession No. XM_291105
Gene Alias ADA2(beta)/ADA2B/MGC21874
Gene Symbols TADA2B
Host Species Mouse
Immunogen MGC21874 (XP_291105, 2 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93624
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.