Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGC87895, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-430 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-430 Supplier Abnova Corporation Supplier No. H00644068K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human MGC87895 peptide using ARM Technology.

This gene is one of the multiple ribosomal protein S14-like genes that are dispersed throughout the genome. This gene is intronless, and may possibly be a pseudogene and/or a null allele of the spliced and functional ribosomal protein S14 gene that is located on chromosome 5. This intronless gene is transcribed and has the potential to encode a protein similar to ribosomal protein S14, but it is unclear as to whether or not a protein is produced. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen MGC87895
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description similar to ribosomal protein S14
Formulation In 1X PBS, pH 7.4
Gene MGC87895
Gene Accession No. MGC87895
Gene Symbols MGC87895
Host Species Rabbit
Immunogen A synthetic peptide of human MGC87895
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644068
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.