Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MgcRacGAP/CYK4 Mouse anti-Human, Clone: 5G5, MilliporeSigma™

Catalog No. MABC588MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC588MI 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC588MI Supplier MilliporeSigma Supplier No. MABC588
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-MgcRacGAP/CYK4 Antibody, clone 5G5 is validated for use in Western Blotting and Immunocytochemistry and Immunoprecipitation for the detection of MgcRacGAP/CYK4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MgcRacGAP/CYK4
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5G5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing PBS without preservatives with 50% glycerol.
Gene Accession No. NP_001119575
Gene Symbols RACGAP1, MGCRACGAP
Host Species Mouse
Immunogen Linear peptide corresponding to Human MgcRacGAP/CYK4.
Purification Method Protein A purified
Quantity 5 g
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.