Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR4 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15097MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15097MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15097MI Supplier MilliporeSigma Supplier No. AB15097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-mGluR4 Antibody is validated for use in western blotting for the detection of mGluR4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.
Gene Symbols GPRC1D ; MGLUR4; mGlu4; mGluR4
Host Species Rabbit
Immunogen Recombinant Protein.
Purification Method Antigen affinity purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001013403.1
Target Species Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.