Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR7 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. 07239MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-239-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-239-MI Supplier MilliporeSigma Supplier No. 07239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect mGluR7 using this Anti-mGluR7 Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols GRM7; GLUR7; mGlu7; mGluR7; MGLUR7; GPRC1G; FLJ40498
Host Species Rabbit
Immunogen peptide (C-NSPAAKKKYVSYNN) corresponding to amino acids 899-912 of human mGluR7
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000831
Target Species Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.