Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR8 Guinea Pig anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB10556MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10556MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10556MI Supplier MilliporeSigma Supplier No. AB10556
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-mGluR8 Antibody detects level of mGluR8 and has been published and validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR8
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified guinea pig polyclonal serum with 0.05% sodium azide.
Gene Symbols GLUR8; mGlu8; GPRC1H; MGLUR8; FLJ41058; MGC126724; GRM8
Host Species Guinea Pig
Immunogen This antibody is generated from guinea pigs immunized with a KLH-conjugated linear peptide selected from residues corresponding to the cytoplasmic domain of rat mGluR8.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_000836.2
Target Species Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.