Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGMT Mouse anti-Human, Clone: MT3.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB16200
Description
Use Anti-MGMT Antibody, clone MT3.1 (mouse monoclonal antibody) validated in flow cytometry, immunocytochemistry,immunohistochemistry (paraffin), immunoprecipitation, western blotting to detect MGMT also known as O-6-methylguanine-DNA Methyltransferase, Methylated-DNA--Protein-cysteine Methyltransferase.
Specifications
MGMT | |
Monoclonal | |
Unconjugated | |
NM_002412.2 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C for one year | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
MT3.1 | |
Purified mouse monoclonal IgG1 in buffer containing 0.02MPB, pH 7.6, 0.25MNaCl containing 0.1% sodium azide. | |
MGMT, EC 2.1.1.63 | |
Made against recombinant human MGMT (O-6-methylguanine-DNA methyltransferase) expressed in E. coli. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction