Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC class II (I-A/I-E) Rat anti-Mouse, Clone: M5/114, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABF33
Description
Detect MHC class II (I-A/I-E) using this Anti-MHC class II (I-A/I-E) Antibody, clone M5/114 validated for use in western blot, flow cytometry, immunohistochemistry.
Specifications
MHC class II (I-A/I-E) | |
Monoclonal | |
Unconjugated | |
NP_001036070 | |
Rat | |
Protein G Purified | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Flow Cytometry, Immunohistochemistry, Western Blot | |
M5/114 | |
Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Ia-GAMMA, DHLAG, CD74, p33, HLADG, ii, Ii | |
B6 Anti-BNL4φ lymphocytes | |
100 μg | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction