Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MHC class II (I-A/I-E), FITC, Clone: M5/114.15.2,

Rat Monoclonal Antibody

$219.36 - $341.12

Specifications

Antigen MHC class II (I-A/I-E)
Clone M5/114.15.2
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF808MI
View Documents
MilliporeSigma™
MABF808
100 μg
Each for $219.36
Only null left
Add to Cart
 
MABF809MI
View Documents
MilliporeSigma™
MABF809
500 μg
Each for $341.12
Only null left
Add to Cart
 
Description

Description

Specifically detects MHC class II (I-A/I-E) Clone: M5/114.15.2 in Mouse samples, and it is validated for Flow Cytometry

The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages, and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.
Specifications

Specifications

MHC class II (I-A/I-E)
Flow Cytometry
FITC
Rat
Inflammation, Immunology
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
CD74, Ii
IgG2b κ
Purified by size-exclusion chromatography
M5/114.15.2
Monoclonal
Purified
RUO
Mouse
P04441
Proprietary immunogen
Primary
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.