Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mi-2Β Mouse anti-Human, Clone: 17H11, MilliporeSigma™

Catalog No. MABE920MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE920MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE920MI Supplier MilliporeSigma Supplier No. MABE920
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Mi-2β Antibody, clone 17H11 is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of Mi-2β.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mi-2β
Applications ChIP Assay, Western Blot
Classification Monoclonal
Clone 17H11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001264
Gene Symbols CHD4
Host Species Mouse
Immunogen Recombinant protein corresponding to Human Mi-2β (CHD-4).
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.