Learn More
MIC A/B anti-Human, Unlabeled, Clone: 6D4, BD
Monoclonal Antibody
Supplier: BD Biosciences 558032
Description
Monoclonal antibody 6D4 reacts with MICA and MICB, distant homologs of major histocompatibility complex class I proteins. They are highly glycosylated molecules of approximately 70kDa that are normally expressed in gut epithelium, are frequently associated with epithelial tumors and can be induced by some viral and bacterial infections. MIC are highly expressed on proliferating but not on quiescent epithelial cell lines, on which they can be heat-shock induced. MIC have similar functional and structural properties and are ligands for NKG2D, an activating receptor on most NK cells, CD8 T cells and ?d T cells. Expression of MIC has been observed on synoviocytes in rheumatoid arthritis and on epithelial cells in transplanted kidney and pancreas showing histological signs of rejection and or cellular injury.
Host Species: Mouse
Clone: 6D4
Isotype: IgG2a, κ
Cross Reactivity: Human
Flow Cytometry

Specifications
MIC A/B | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
6D4 | |
Unconjugated | |
MICA transfected cells | |
0.1 mg | |
Cell Biology | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.