Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICB, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005765 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-765 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-765 Supplier Abnova Corporation Supplier No. H00004277B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MICB protein.

This gene encodes a heavily glycosylated protein which is a ligand for the NKG2D type II receptor. Binding of the ligand activates the cytolytic response of natural killer (NK) cells, CD8 alphabeta T cells, and gammadelta T cells which express the receptor. This protein is stress-induced and is similar to MHC class I molecules; however, it does not associate with beta-2-microglobulin or bind peptides. [provided by RefSeq

Sequence: MGLGRVLLFLAVAFPFAPPAAAAEPHSLRYNLMVLSQDGSVQSGFLAEGHLDGQPFLRYDRQKRRAKPQGQWAEDVLGAETWDTETEDLTENGQDLRRTLTHIKDQKGVPQSSRAQTLAMNVTNFWKEDAMKTKTHYRAMQADCLQKLQRYLKSGVAIRRTVPPMVNVTCSEVSEGNITVTCRASSFYPRNITLTWRQDGVSLSHNTQQWGDVLPDGNGTYQTWVATRIRQGEEQRFTCYMEHSGNHGTHPVPSGKALVLQSQRTDFPYVSAAMPCFVIIIILCVPCCKKKTSAAEGPELVSLQVLDQHPVGTGDHRDAAQLGFQPLMSATGSTGSTEGA

Specifications

Antigen MICB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MICB protein.
Formulation No additive
Gene MICB
Gene Accession No. BC044218.1
Gene Alias PERB11.2
Gene Symbols MICB
Host Species Mouse
Immunogen MICB (AAH44218.1, 1 a.a. ∼ 340 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4277
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.