Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microglia Mouse anti-Human, Clone: 25F9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1569
Description
Anti-Microglia Antibody, clone 25F9 detects level of Microglia and has been published and validated for use in flow cytometry, immunocytochemistry, immunohistochemistry.
Specifications
Microglia | |
Monoclonal | |
Unconjugated | |
Mouse | |
Primary | |
−20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
25F9 | |
Purified immunoglobulin from culture supernatant. Lyophilized from PBS, pH 7.2 containing 5mg/mL BSA and 0.09% sodium azide. Reconstitute with 500μL of sterile distilled water. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction