Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microphthalmia (Mi) Mouse anti-Human, Rat, Mouse, Clone: C5, MilliporeSigma™

Catalog No. MAB3747MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3747MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3747MI Supplier MilliporeSigma Supplier No. MAB3747
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Microphthalmia (Mi) Antibody, clone C5 is a Mouse Monoclonal Antibody for detection of Microphthalmia (Mi) also known as Microphthalmia-associated transcription factor and has been tested in EMSA, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Microphthalmia (Mi)
Applications Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in phosphate buffered saline with 0.08% sodium azide.
Gene Symbols LZMS, MCOPS1, MAA
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from RBF/DnJ mice immunized with an N-terminal fragment of human microphthalmia protein and mouse myeloma NS1 cells.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.