Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Midkine Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA528823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528823 Supplier Thermo Scientific Supplier No. PA528823

Rabbit Polyclonal Antibody

PA5-28823 targets Midkine in WB applications and shows reactivity with Human samples. The PA5-28823 immunogen is synthetic peptide corresponding to a region within amino acids 79 and 143 of Human Midkine. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis (summary by Reynolds et al., 2004 [PubMed 15197188]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Midkine
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Gene MDK
Gene Accession No. P21741
Gene Alias MK, NEGF2, ARAP, amphiregulin-associated protein, midgestation and kidney protein, midkine, neurite outgrowth-promoting factor 2, neurite outgrowth-promoting protein
Gene Symbols MDK
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 79 and 143 of Human Midkine
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4192
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.