Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIG Mouse anti-Human, Biotin, Clone: B8-6, BD

Catalog No. BDB555037 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555037 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555037 Supplier BD Biosciences Supplier No. 555037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody B8-6 reacts with human monokine induced by gamma interferon (MIG). MIG is inducible in macrophages, hepatocytes, and endothelial cells by IFN-γ, but not by IFN-α or bacterial lipopolysaccharides. The immunogen used to generate the monoclonal antibody B8-6 was insect cell-expressed, recombinant human MIG protein.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIG
Applications ELISA
Classification Monoclonal
Clone B8-6
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human MIG Protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.