Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mint1 Mouse, Unlabeled, Clone: 23, BD

Catalog No. BDB611029 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611029 150 μg
BDB611028 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611029 Supplier BD Biosciences Supplier No. 611029

Mouse Monoclonal Antibody

Neuronal communication via neurotransmitter release is mediated by the synaptic vesicle cycle. The initial step in exocytosis is the docking of the vesicle in the active zone of the plasma membrane. This step is followed by fusion of the vesicle and plasma membrane and exocytosis. Munc18-1, a major brain protein, is essential for exocytosis. It binds the vesicle fusion protein syntaxin, along with Doc2a and 2b, two proteins that associate peripherally with the synaptic vesicle. Munc18-1 is a family member of membrane trafficking proteins. Its function is thought to be mediated by two Munc18-1-interacting proteins termed Mint 1 and Mint 2, which are 50% homologous. They are expressed exclusively in brain and bind Munc18-1 (MID) with high affinity. They contain an N-terminal Munc18-1 interacting domain and C-terminal PTB (pTyr/PIP interaction) and PDZ (membrane protein interaction) domains, suggesting the ability of Mint proteins to link vesicle exocytosis to Tyr phosphorylation and/or localization at synaptic intercellular junctions. Thus, the Mint proteins, along with Munc18-1 and syntaxin, form a multimeric complex that mediates appropriate docking/fusion of synaptic vesicles.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mint1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat Mint 1 aa. 268-377
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rabbit, Rat
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.