Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIP-1α Mouse anti-Human, APC, Clone: 11A3, BD

Catalog No. BDB551533 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551533 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551533 Supplier BD Biosciences Supplier No. 551533
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The immunogen used to raise R139 was a CDw93 preparation as described. CDw93 is also known as C1q-binding protein. Human C1qRp is a 631 amino acid protein (∽66.5kD) protein that is highly expressed on monocytes/macrophages, neutrophil granulocytes but not on T and B lymphocytes. C1qRp binds C1q, the recognition subunit of the first component (C1) of the complement pathway, as well as MBL (Mannose-binding-lectin) and SPA(Pulmonary Surfactant Protein A). Human C1qRp is involved in the C1q-mediated enhancement of phagocytosis. R139 is suitable to detect C1qRp expression on cells of myeloid lineage by flow cytometry, C1qRp in cellular lysates by Western blotting or immunoprecipitation. In addition, R139 neutralizes C1qmediated enhancement of phagocytosis, as reported. CDw93 (C1qRp) has been reported to define a human stem cell population with hematopoietic and hepatic potential.

Host Species: Mouse
Clone: 11A3
Isotype: IgG2b κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1α
Applications Flow Cytometry
Classification Monoclonal
Clone 11A3
Concentration 0.5mg/mL
Conjugate APC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.