Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIP-1β Mouse anti-Human, FITC, Clone: D21-1351, BD

Catalog No. BDB560565 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560565 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560565 Supplier BD Biosciences Supplier No. 560565
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody D21-1351 reacts with the human CC chemokine, MIP-1β (macrophage inflammatory protein-1β). Human MIP-1β shares approximately 75% homology with mouse MIP-1β at the amino acid level. Expression of MIP-1β in hu

Host Species: Mouse
Clone: D21-1351
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Recombinant Human MIP-1β

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1β
Applications Flow Cytometry
Classification Monoclonal
Clone D21-1351
Conjugate FITC
Description Macrophage inflammatory protein 1-beta; CCL4; C-C motif chemokine 4; LAG-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human MIP-1β
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.