Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIP-1α Mouse anti-Human, PE, Clone: 11A3, BD

Catalog No. BDB554730 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554730 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554730 Supplier BD Biosciences Supplier No. 554730
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with macrophage inflammatory protein-1α (MIP-1α), a member of the CC chemokine family of proteins. It is produced by T cells, B cells, Langerhans cells, neutrophils and macrophages. MIP-1α plays a role as an inhibitor of stem cell proliferation and as a chempattractant of B cells, eosinophils and cytotoxic T cells. Clone 11A3 also cross-reacts with an intracellular component of rhesus and cynomolgus macaque-LPS-stimulated peripheral blood monocytes. The reactivity pattern observed on CD14-positive cells is similar to that seen on normal human peripheral blood monocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.

Host Species: Mouse
Clone: 11A3
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Mouse IgG2a, κ

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1α
Applications Flow Cytometry
Classification Monoclonal
Clone 11A3
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen IgG2a, κ
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.