Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MiTF Mouse anti-Human, Mouse, Clone: 3D1.2, MilliporeSigma™

Catalog No. MABE78MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE78MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE78MI Supplier MilliporeSigma Supplier No. MABE78
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MiTF Antibody, clone 3D1.2 is an antibody against MiTF for use in Western Blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MiTF
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3D1.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000239
Gene Symbols MITF, BHLHE32
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human MiTF.
Purification Method Purified by ion-exchange chromatography
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.