Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitochondria Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™

Catalog No. AB3598MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3598MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3598MI Supplier MilliporeSigma Supplier No. AB3598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Mitochondria Antibody is an antibody against Mitochondria for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitochondria
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum preabsorbed against immunogen peptide. Liquid. Contains 0.01% sodium azide.
Gene Symbols PSEN1; PS1; PSNL1; PS-1; AD3; FAD; S182; Presenilin-1; EC 3.4.23.- [Contains: Presenilin-1 NTF subunit; Presenilin-1 CTF subunit; Presenilin-1 CTF12 (PS1-CTF12)].
Host Species Rabbit
Immunogen Presenilin 1 peptide conjugated to KLH.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000021.2
Target Species Human, Monkey
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.