Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP kinase interacting serine/threonine kinase 2, Mouse, Clone: 2A10, Abnova™

Catalog No. 89000695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-695 Supplier Abnova Corporation Supplier No. H00002872M07
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MKNK2.

Sequence: MAPEVVEAFSEEASIYDKRCDLWSLGVILYILLSGYPPFVGRCGSDCGWDRGEACPACQNMLFESIQEGKYEFPDKDWAHISCAAKDLISKLLVRDAKQRLSAAQVLQHPWVQGCAPENTLPTPMVLQRWDSHFLLPPHPCRIHVRPGGLVRTVTVNE

Specifications

Antigen MAP kinase interacting serine/threonine kinase 2
Applications ELISA, In situ PLA, Western Blot
Classification Monoclonal
Clone 2A10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MKNK2.
Formulation PBS with no preservative; pH 7.4
Gene MKNK2
Gene Accession No. BC018345
Gene Alias GPRK7/MNK2
Gene Symbols MKNK2
Host Species Mouse
Immunogen MKNK2 (AAH18345, 1 a.a. ∼ 158 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2872
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.