Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKP4, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127644
Description
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regu
PA1-27644 detects MKP4 from human samples.PA1-27644 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27913 immunogen is a synthetic peptide corresponding to human Dual specificity protein phosphatase homolog hMKP-R.
Specifications
| MKP4 | |
| Polyclonal | |
| DUSP9 | |
| MKP-4, MKP4 | |
| Chicken | |
| Antigen affinity chromatography | |
| RUO | |
| 1852 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Q99956 | |
| DUSP9 | |
| Synthetic peptide corresponding to human Dual specificity protein phosphatase homolog hMKP-R. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction