Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MLH1 Clone: 14,

Catalog No. NA28100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NA28100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NA28100UG Supplier MilliporeSigma™ Supplier No. NA28100UG

Mouse Monoclonal Antibody

Specifically detects MLH1 Clone: 14 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

The MLH1 gene is one of at least 4 genes encoding proteins involved in the repair of mismatch nucleotides following DNA replication or repair. Mutations in the MLH1 gene contribute to the development of sporadic colorectal carcinoma, while germline MLH1 mutations are responsible for ∽35% of inherited, non-polyposis colorectal carcinoma (HNPCC). MLH1 encodes a protein of 756 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLH1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 14
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen recombinant, human MLH1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽83kDa MLH1 protein in LoVo cells.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.