Learn More
MLH1 Mouse anti-Human, Unlabeled, Clone: G168-728, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 554073
Description
The repair of mismatched DNA is essential to maintaining the integrity of genetic information over time. Loss of function of DNA repair enzymes can lead to an accumulation of replication errors, resulting in a mutated phenotype. DNA repair enzymes are highly conserved from bacteria to yeast to mammals. In yeast the proteins are called MutS homolog 2 (MSH2), MutL homolog (MLH1), and PMS1 which is also a homolog of MutL. MSH2 is involved in the initial recognition of mismatched nucleotides during the replication mismatch repair process. It is thought that after MSH2 binds to a mismatched DNA duplex, it is joined by a heterodimer of MLH1 and PMS1 which together help facilitate the later steps in mismatch repair. The G168-728 antibody recognizes human and mouse MLH1 (80-85kDa). Full-length human recombinant MLH was expressed as a maltose binding-MLH fusion protein, affinity purified, and used as immunogen.
Host Species: Mouse
Clone: G168-728
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Recombinant Human MLH
Formula Weight [Chemical]: 80-85kDa
Immunoprecipitation, Western Blotting

Specifications
MLH1 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Human MLH | |
0.1 mg | |
Cell Biology | |
Human, Mouse | |
IgG2a κ |
Western Blot | |
G168-728 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.