Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLL/HRX Mouse anti-Human, Mouse, Clone: 9-12, MilliporeSigma™

Catalog No. 05765MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-765-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-765-MI Supplier MilliporeSigma Supplier No. 05765
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MLL/HRX, C-term., clone 9-12 is a high quality mouse monoclonal antibody that has been validated and published for use in Chromatin Immunoprecipitation (ChIP), Immunofluorescence, Immunoprecipitation, and Western Blotting applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLL/HRX
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9-12
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative.
Gene Accession No. NM_005933
Gene Symbols MLL, HRX, CXXC7, ALL1, HTRX1, TRX1, FLJ11783, ALL-1, MLL/GAS7, HTRX, MLL1A
Host Species Mouse
Immunogen MBP fusion protein corresponding to residues 3084-3959 of human mixed lineage leukemia (MLL)
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.