Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6, Mouse, Clone: 1B2, Abnova™

Catalog No. 89000809 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-809 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-809 Supplier Abnova Corporation Supplier No. H00004302M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MLLT6.

Sequence: MGAVNPLLSQAESSHTEPDLEDCSFRCRGTSPQESLSSMSPISSLPALFDQTASAPCGGGQLDPAAPGTTNMEQLLEKQGDGEAGVNIVEMLKALHALQKENQRLQEQILSLTAKKERLQILNVQLSVPFPALPAALPAANGPVPGPYGLPPQAGSSDSLSTSKSPPGKSSLGLDNSLSTSSEDPHSGCPSRSSSSLSFHSTPPPLPLLQQSPATLPLALPGAPAPLPPQPQNGLGRAPGAAGLGAMPMAEGLLGGLAGSGGLPLNGLLGGLNGAAAPNPASLSQAGGAPTLQLPGCLNSLTEQQRHLLQQQEQQLQQLQQLLASPQLTPEHQTVVYQMIQQIQQKRELQRLQMAGGSQLPMASLLAGSSTPLLSAGTPGLLPTASAPPLLPAGALVAPSLGNNTSLMAAAAAAAAVAAAGGPPVLTAQTNPFLSLSGAEGSGGGPKGGTADKGASANQEKG

Specifications

Antigen myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MLLT6.
Formulation PBS with no preservative; pH 7.4
Gene MLLT6
Gene Accession No. BC007237
Gene Alias AF17/FLJ23480
Gene Symbols MLLT6
Host Species Mouse
Immunogen MLLT6 (AAH07237, 1 a.a. ∼ 462 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4302
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.