Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-1 Mouse anti-Human, Clone: 2A7.2, MilliporeSigma™

Catalog No. MAB13438MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB13438MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB13438MI Supplier MilliporeSigma Supplier No. MAB13438

Mouse Monoclonal Antibody

Anti-MMP-1 Antibody, clone 2A7.2 is an antibody against MMP-1 for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A7.2
Conjugate Unconjugated
Formulation Liquid at 1mg/mL containing 0.1% sodium azide.
Gene Accession No. NM_002421.2
Gene Symbols MMP1, CLG, MMP-1, CLGN, EC 3.4.24.7 [Contains: 22 kDa interstitial collagenase, 27 kDa interstitial collagenase].
Host Species Mouse
Immunogen Synthetic peptide corresponding to the amino terminal region of human MMP-1.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.