Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-14 Mouse anti-Mouse, Clone: LEM-2/15.8, MilliporeSigma™

Catalog No. MAB3328MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3328MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3328MI Supplier MilliporeSigma Supplier No. MAB3328
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect MMP-14 using this Anti-MMP-14 Antibody, catalytic domain, clone LEM-2/15.8 validated for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-14
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LEM-2/15.8
Conjugate Unconjugated
Formulation Purified immunoglobulin by Protein A chromatography . Liquid in 0.2M phosphate, 0.25M NaCl, pH 7.6, containing 0.1% sodium azide.
Gene Accession No. NM_004995.2
Gene Symbols MMP14, MTMMP1, MMP-14, MT1MMP, MT1-MMP, MMP-X1, EC 3.4.24.80
Host Species Mouse
Immunogen Synthetic peptide: amino acid sequence 218-233 within the catalytic domain
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.